missing translation for 'onlineSavingsMsg'
Learn More

1-Acetonylpyridinium Chloride 99.0+%, TCI America™

Catalog No. A08285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A08285G Supplier TCI America Supplier No. A08285G

Specifications

CAS 42508-60-1
Melting Point 207°C
Color White-Yellow
Molecular Formula C8H10ClNO
Quantity 5 g
MDL Number MFCD00031949
Synonym 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride
InChI Key JHYYWKUENNZTMD-UHFFFAOYSA-M
SMILES [Cl-].CC(=O)C[N+]1=CC=CC=C1
IUPAC Name 1-(2-oxopropyl)pyridin-1-ium chloride
Molecular Weight (g/mol) 171.62
PubChem CID 2734121
Formula Weight 171.62
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetonylpyridinium Chloride
Show More Show Less