missing translation for 'onlineSavingsMsg'
Learn More

1,9-Decadiene 98.0+%, TCI America™

Catalog No. D02455ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02455ML 5 mL
D024525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02455ML Supplier TCI America Supplier No. D02455ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1647-16-1
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00008670
InChI Key NLDGJRWPPOSWLC-UHFFFAOYSA-N
Synonym 1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020
PubChem CID 15439
IUPAC Name deca-1,9-diene
SMILES C=CCCCCCCC=C

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 mL
UN Number 3295
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,9-Decadiene
EINECSNumber (2)-3625
TSCA Yes