missing translation for 'onlineSavingsMsg'
Learn More

1,8-Nonadiene 98.0+%, TCI America™

Catalog No. p-7133754
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03325ML 5 mL
N033225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03325ML Supplier TCI America Supplier No. N03325ML

Chemical Identifiers

CAS 4900-30-5
Molecular Formula C9H16
Molecular Weight (g/mol) 124.227
MDL Number MFCD00008669
InChI Key VJHGSLHHMIELQD-UHFFFAOYSA-N
Synonym 1,8-nonadiene, acmc-209kep
PubChem CID 78612
IUPAC Name nona-1,8-diene
SMILES C=CCCCCCC=C

Specifications

Color Colorless
Boiling Point 132°C
Quantity 5 mL
UN Number 3295
Formula Weight 124.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Nonadiene