missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Naphthyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 98°C |
| Quantity | 1 g |
| Formula Weight | 130.15 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,8-Naphthyridine |
Safety and Handling
| TSCA | No |