missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxyanthraquinone, 95%

Catalog No. AAA1200136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1200136 500 g
AAA1200122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1200136 Supplier Thermo Scientific Chemicals Supplier No. A1200136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 117-10-2 | C14H8O4 | 240.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117-10-2
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001211
InChI Key QBPFLULOKWLNNW-UHFFFAOYSA-N
Synonym danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
PubChem CID 2950
ChEBI CHEBI:3682
IUPAC Name 1,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O

Specifications

Melting Point 191°C to 193°C (sublimation)
Quantity 500 g
Beilstein 2054727
Merck Index 14,2815
Formula Weight 240.21
Percent Purity 95%
Chemical Name or Material 1,8-Dihydroxyanthraquinone
Hazard Category H319-H350
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 204-173-5
RTECSNumber CB6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only