missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Diaminooctane, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 373-44-4 |
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Density | 165g/mL |
| Boiling Point | 225.0°C to 226.0°C |
| Flash Point | 115°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C8H22Cl2N2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. Wear |
| EINECSNumber | 206-764-3 |
RUO – Research Use Only