missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane 97.0+%, TCI America™

Catalog No. D52675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52675G 5 g
D526725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52675G Supplier TCI America Supplier No. D52675G

Chemical Identifiers

CAS 2474-02-4
Molecular Formula C8H24Cl2O3Si4
Molecular Weight (g/mol) 351.52
MDL Number MFCD00045148
InChI Key UHRAUGIQJXURFE-UHFFFAOYSA-N
PubChem CID 75584
IUPAC Name chloro-[[[chloro(dimethyl)silyl]oxy-dimethylsilyl]oxy-dimethylsilyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)O[Si](C)(C)Cl)O[Si](C)(C)Cl

Specifications

Melting Point -62°C
Color Colorless
Boiling Point 222°C
Quantity 5 g
UN Number 2987
Formula Weight 351.52
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane
EINECSNumber (7)-0472
TSCA Yes