missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol 97.0+%, TCI America™

Catalog No. H0099500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0099500G 500 g
H009925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0099500G Supplier TCI America Supplier No. H0099500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-11-8
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00002985
InChI Key XXMIOPMDWAUFGU-UHFFFAOYSA-N
Synonym 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol
PubChem CID 12374
ChEBI CHEBI:43078
IUPAC Name hexane-1,6-diol
SMILES C(CCCO)CCO

Specifications

Melting Point 42°C
Color White
Boiling Point 208°C
Quantity 500 g
Formula Weight 118.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Hexanediol
EINECSNumber (2)-0240
RTECSNumber MO2100000
TSCA Yes