missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadiyne 98.0+%, TCI America™

Catalog No. H04835ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04835ML 5 mL
H04831ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04835ML Supplier TCI America Supplier No. H04835ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2396-63-6
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
MDL Number MFCD00014925
InChI Key RSPZSDWVQWRAEF-UHFFFAOYSA-N
Synonym 1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
PubChem CID 337121
IUPAC Name hepta-1,6-diyne
SMILES C#CCCCC#C

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 mL
UN Number 3295
Formula Weight 92.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Heptadiyne
RTECSNumber MI5600000
TSCA Yes