missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromopyrene 98.0+%, TCI America™

Catalog No. D31675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31675G 5 g
D31671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31675G Supplier TCI America Supplier No. D31675G

Chemical Identifiers

CAS 27973-29-1
Molecular Formula C16H8Br2
Molecular Weight (g/mol) 360.048
MDL Number MFCD09263746
InChI Key JRCJYPMNBNNCFE-UHFFFAOYSA-N
PubChem CID 176470
IUPAC Name 1,6-dibromopyrene
SMILES C1=CC2=C(C=CC3=C2C4=C1C=CC(=C4C=C3)Br)Br

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 5 g
Formula Weight 360.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Dibromopyrene