missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™

Catalog No. D20195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20195G 5 g
D201925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20195G Supplier TCI America Supplier No. D20195G
Only null left

Chemical Identifiers

CAS 1633-00-7
Molecular Formula C14H24N2O8
Molecular Weight (g/mol) 348.35
MDL Number MFCD00004292
InChI Key YGDVXSDNEFDTGV-UHFFFAOYSA-N
Synonym 1,6-diaminohexane-n,n,n',n'-tetraacetic acid, hexamethylenediaminetetraacetic acid, hexamethylenediamine-n,n,n',n'-tetraacetic acid, 2-6-bis carboxymethyl amino hexyl carboxymethyl amino acetic acid, hdta, glycine,n,n'-1,6-hexanediylbis n-carboxymethyl, 6-bis carboxymethyl amino hexyl carboxymethyl amino acetic acid, glycine, n,n'-1,6-hexanediylbis n-carboxymethyl, 1,6-hexanediaminetetraacetic acid, 2,2',2,2'-hexane-1,6-diyldinitrilo tetraacetic acid
PubChem CID 74209
IUPAC Name 2-({6-[bis(carboxymethyl)amino]hexyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCCCCCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point 236°C
Color White
Quantity 5 g
Formula Weight 348.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Diaminohexane-N,N,N′,N′-tetraacetic Acid