missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-beta-D-glucose 99.0+%, TCI America™

Catalog No. A10745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10745G 5 g
A10741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10745G Supplier TCI America Supplier No. A10745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 498-07-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00063248
InChI Key TWNIBLMWSKIRAT-UHFFFAOYNA-N
Synonym 1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucose
PubChem CID 2724705
ChEBI CHEBI:30997
IUPAC Name 6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
SMILES OC1C2COC(O2)C(O)C1O

Specifications

Melting Point 183°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Anhydro-beta-D-glucose