missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-beta-D-glucopyranose, 99%

Catalog No. AAA1672403
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1672403 1 g
AAA1672401 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1672403 Supplier Thermo Scientific Chemicals Supplier No. A1672403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 498-07-7 | C6H10O5 | 162.14 g/mol

1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

Solubility
Soluble in Ethanol and Water.

Notes
Store at -20°C. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 498-07-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00063248
InChI Key TWNIBLMWSKIRAT-UHFFFAOYNA-N
Synonym 1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucose
PubChem CID 2724705
ChEBI CHEBI:30997
IUPAC Name (1R,2S,3S,4R,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
SMILES OC1C2COC(O2)C(O)C1O

Specifications

Melting Point 180°C to 184°C
Quantity 1 g
Beilstein 80998
Solubility Information Soluble in Ethanol and Water.
Formula Weight 162.14
Percent Purity 99%
Chemical Name or Material 1,6-Anhydro-beta-D-glucopyranose
EINECSNumber 207-855-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only