missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,6-Anhydro-beta-D-cellobiose, 95%

Catalog No. AAJ66914ME
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66914ME 500 mg
AAJ6691403 1 g
AAJ6691406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66914ME Supplier Thermo Scientific Chemicals Supplier No. J66914ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35405-71-1 | C12H20O10 | 324.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35405-71-1
Molecular Formula C12H20O10
Molecular Weight (g/mol) 324.282
MDL Number MFCD13182343
InChI Key LTYZUJSCZCPGHH-QRZGKKJRSA-N
Synonym 1,6-anhydro-b-d-cellobiose, cellobiosan, 1,6-anhydro-beta-d-cellobiose, 2s,3r,4s,5s,6r-2-1r,2s,3r,4r,5r-3,4-dihydroxy-6,8-dioxabicyclo 3.2.1 octan-2-yl oxy-6-hydroxymethyl oxane-3,4,5-triol
PubChem CID 71225847
IUPAC Name (2S,3R,4S,5S,6R)-2-[[(1R,2S,3R,4R,5R)-3,4-dihydroxy-6,8-dioxabicyclo[3.2.1]octan-2-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C2C(C(C(C(O1)O2)O)O)OC3C(C(C(C(O3)CO)O)O)O

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water at 50mg/ml
Formula Weight 324.28
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1,6-Anhydro-beta-D-cellobiose
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only