missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride, 95%

Catalog No. AAH34430MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34430MD 250 mg
AAH3443003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34430MD Supplier Thermo Scientific Chemicals Supplier No. H34430MD

CAS: 1006441-36-6 | C9H5ClF3N3O2S | 311.663 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1006441-36-6
Molecular Formula C9H5ClF3N3O2S
Molecular Weight (g/mol) 311.663
MDL Number MFCD08689729
InChI Key ZACLAYCACGRQPN-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-5-trifluoromethyl-2-pyridinyl-1h-pyrazole-4-sulfonoyl chloride, 1-5-trifluoromethyl pyridin-2-yl pyrazole-4-sulfonyl chloride, 1-5-trifluoromethyl pyridin-2-yl-1h-pyrazole-4-sulphonyl chloride, trifluoromethylpyridinylpyrazolesulfonoylchloride, chloro 1-5-trifluoromethyl 2-pyridyl pyrazol-4-yl sulfone, 1-5-trifluoromethyl pyridin-2-yl-1h-pyrazole-4-sulfonylchloride
PubChem CID 18526157
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]pyrazole-4-sulfonyl chloride
SMILES C1=CC(=NC=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl

Specifications

Melting Point 87°C to 89°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 311.67
Percent Purity 95%
Chemical Name or Material 1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only