missing translation for 'onlineSavingsMsg'
Learn More

1,5-Hexadiene Diepoxide 96.0+%, TCI America™

Catalog No. H09531G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09531G 1 g
H09535G 5 g
H095325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H09531G Supplier TCI America Supplier No. H09531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1888-89-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD01723447
InChI Key HTJFSXYVAKSPNF-UHFFFAOYSA-N
Synonym 1,5-hexadiene diepoxide, 1,2,5,6-diepoxyhexane, 1,2:5,6-diepoxyhexane, 2-2-oxiran-2-yl ethyl oxirane, ccris 3751, hexane, 1,2:5,6-diepoxy, 1,2-di oxiran-2-yl ethane, 1,6-diepoxyhexane, 1,2-bisoxiranylethane, 1,5,6-diepoxyhexane
PubChem CID 15903
IUPAC Name 2-[2-(oxiran-2-yl)ethyl]oxirane
SMILES C1C(O1)CCC2CO2

Specifications

Color Yellow
Boiling Point 188°C
Quantity 1 g
Formula Weight 114.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Hexadiene Diepoxide
RTECSNumber MO1575000
TSCA No