missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Fluoro-2-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Numéro de catalogue. BTB04130EE
Modifier l'affichage
Click to view available options
Quantité:
50g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
BTB04130EE 50g
BTB04130FA 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. BTB04130EE Fournisseur Thermo Scientific Chemicals Code fournisseur BTB04130EE

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 394-32-1
Formule moléculaire C8H7FO2
Poids moléculaire (g/mol) 154.14
Clé InChI KOFFXZYMDLWRHX-UHFFFAOYSA-N
Synonyme 5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone
PubChem CID 95992
Nom de l’IUPAC 1-(5-fluoro-2-hydroxyphenyl)ethanone
SOURIRES CC(=O)C1=C(C=CC(=C1)F)O

Spécifications

Nom ou substance chimique 1-(5-Fluoro-2-hydroxyphenyl)ethan-1-one
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule linéaire FC6H3(OH)COCH3
Quantité 50g
Poids de formule 154.14
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.