missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dinitronaphthalene 98.0+%, TCI America™
Supplier: TCI America D083625G
Specifications
| 1,5-Dinitronaphthalene | |
| 218°C | |
| C10H6N2O4 | |
| 25 g | |
| ZUTCJXFCHHDFJS-UHFFFAOYSA-N | |
| 1,5-dinitronaphthalene | |
| 11803 | |
| 218.17 | |
| Crystalline Powder |
| 605-71-0 | |
| Yellow | |
| MFCD00003916 | |
| naphthalene, 1,5-dinitro, naphthalene, dinitro, unii-ui2s14iw4t, ccris 6028, ui2s14iw4t, napththalene, 1,5-dinitro, 1,5 or 1,8-dinitronaphthalene, acmc-1bixi, 1,5-dinitro naphthalene, 1,5-dinitro-naphthalene | |
| C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)[N+](=O)[O-] | |
| 218.168 | |
| CHEBI:50640 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 605-71-0 | |
| 218.168 | |
| ZUTCJXFCHHDFJS-UHFFFAOYSA-N | |
| 11803 | |
| 1,5-dinitronaphthalene |
| C10H6N2O4 | |
| MFCD00003916 | |
| naphthalene, 1,5-dinitro, naphthalene, dinitro, unii-ui2s14iw4t, ccris 6028, ui2s14iw4t, napththalene, 1,5-dinitro, 1,5 or 1,8-dinitronaphthalene, acmc-1bixi, 1,5-dinitro naphthalene, 1,5-dinitro-naphthalene | |
| CHEBI:50640 | |
| C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)[N+](=O)[O-] |
Safety and Handling
EINECSNumber : (4)-0325
RTECSNumber : QJ4551000
TSCA : No