accessibility menu, dialog, popup

UserName

ONONOO

CAS RN 605-71-0

IUPAC Name: 1,5-dinitronaphthalene
Synonyms: 1,5-dinitronaphthalene
Molecular Weight (g/mol): 218.17
Molecular Formula: C10H6N2O4
InChi Key: ZUTCJXFCHHDFJS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C(C=CC=C12)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)