missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dinitronaphthalene 98.0+%, TCI America™

Catalog No. p-7126461
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D083625G 25 g
D0836250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D083625G Supplier TCI America Supplier No. D083625G
Only null left

Chemical Identifiers

CAS 605-71-0
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.168
MDL Number MFCD00003916
InChI Key ZUTCJXFCHHDFJS-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dinitro, naphthalene, dinitro, unii-ui2s14iw4t, ccris 6028, ui2s14iw4t, napththalene, 1,5-dinitro, 1,5 or 1,8-dinitronaphthalene, acmc-1bixi, 1,5-dinitro naphthalene, 1,5-dinitro-naphthalene
PubChem CID 11803
ChEBI CHEBI:50640
IUPAC Name 1,5-dinitronaphthalene
SMILES C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)[N+](=O)[O-]

Specifications

Melting Point 218°C
Color Yellow
Quantity 25 g
Formula Weight 218.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dinitronaphthalene
EINECSNumber (4)-0325
RTECSNumber QJ4551000
TSCA No