missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diiodo-2,4-diméthylbenzène 97,0+%, TCI America™

Catalog No. D50055G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
D50055G 5 g
D50051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50055G Supplier TCI America Supplier No. D50055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4102-50-5 | C8H8I2 | 357.96 g/mol

Chemical Identifiers

CAS 4102-50-5
Formule moléculaire C8H8I2
Poids moléculaire (g/mol) 357.96
Numéro MDL MFCD29089354
Clé InChI ZOOZGIAPEXFLEF-UHFFFAOYSA-N
Synonyme 4,6-Diiodo-m-xylene
PubChem CID 12575960
Nom de l’IUPAC 1,5-diiodo-2,4-diméthylbenzène
SOURIRES CC1=CC(C)=C(I)C=C1I

Specifications

Nom ou substance chimique 1,5-Diiodo-2,4-dimethylbenzene
Point de fusion 72 °C
Quantité 5 g
Emballage Glass Bottle
Forme physique Poudre de cristal à 20 °C
Poids de formule 357.96
Pourcentage de pureté ≥97,0% (GC)
Niveau Not Available