missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminonaphthalene, Matrix Substance for MALDI-MS, 99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018764
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018764 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018764 Supplier Merck Emd Millipore Supplier No. 56451250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1,5-Diaminonaphthalene is used in the production of 1,5-naphthalene diisocyanate and polyurethane elastomers.

Specifications

CAS 2243-62-1
Melting Point 185°C to 187°C
Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C10H6(NH2)2
Quantity 250 mg
MDL Number MFCD00004029
Synonym 1,5-DAN; 1,5-Naphthalenediamine; NSC 401110
Formula Weight 158.20
Percent Purity ≥99.0% (HPLC)
Grade Matrix Substance for MALDI-MS