Learn More
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 1215564-15-0 |
|---|---|
| Molecular Formula | C8H4ClF3N4O2S |
| Molecular Weight (g/mol) | 312.651 |
| MDL Number | MFCD11505058 |
| InChI Key | SADUDBJMXGGXDE-UHFFFAOYSA-N |
| Synonym | 1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone |
| PubChem CID | 51063957 |
| IUPAC Name | 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride |
| SMILES | C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH33424MD
|
Thermo Scientific Chemicals
H33424MD |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAH3342403
|
Thermo Scientific Chemicals
H3342403 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1215564-15-0 | |
| 312.651 | |
| SADUDBJMXGGXDE-UHFFFAOYSA-N | |
| 51063957 | |
| C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl |
| C8H4ClF3N4O2S | |
| MFCD11505058 | |
| 1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone | |
| 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride |
Specifications
| 1215564-15-0 | |
| MFCD11505058 | |
| UN3261 | |
| 1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone | |
| C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl | |
| 312.651 | |
| 312.66 | |
| 1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride |
| C8H4ClF3N4O2S | |
| 250mg | |
| Moisture sensitive | |
| SADUDBJMXGGXDE-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride | |
| 51063957 | |
| 95% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO â Research Use Only