missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Numéro de catalogue. p-2694059
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC00119DA 1g
CC00119DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC00119DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC00119DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 423768-49-4
Formule moléculaire C11H8ClFN2O
Poids moléculaire (g/mol) 238.646
Numéro MDL MFCD03659694
Clé InChI INIAJXQDJISNJB-UHFFFAOYSA-N
Synonyme 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methyl-4-pyrazolecarbonyl chloride, 1-4-fluorophenyl-5-methyl-pyrazole-4-carbonyl chloride
PubChem CID 2779893
Nom de l’IUPAC 1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride
SOURIRES CC1=C(C=NN1C2=CC=C(C=C2)F)C(=O)Cl

Spécifications

Nom ou substance chimique 1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 238.65
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.