missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile 98.0+%, TCI America™

Catalog No. F08791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08791G Supplier TCI America Supplier No. F08791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 64169-67-1
Melting Point 100°C
Color Yellow
Molecular Formula C15H10FNO
Quantity 1 g
MDL Number MFCD06200812
InChI Key YXCRMKYHFFMNPT-UHFFFAOYNA-N
SMILES FC1=CC=C(C=C1)C1OCC2=C1C=CC(=C2)C#N
IUPAC Name 1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile
Molecular Weight (g/mol) 239.25
PubChem CID 9856159
Formula Weight 239.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile
Show More Show Less
TSCA No
Recommended Storage Freezer