missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dipropylbenzene 97.0+%, TCI America™

Supplier:  TCI America D278025ML

 View more versions of this product

Catalog No. D278025ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,4-Dipropylbenzene
Yellow
C12H18
25 mL
CCCC1=CC=C(C=C1)CCC
162.276
162.28
Liquid
4815-57-0
221°C
MFCD03844789
PHUANMGFAOCUOQ-UHFFFAOYSA-N
1,4-dipropylbenzene
20958
≥97.0% (GC)

Chemical Identifiers

4815-57-0
162.276
PHUANMGFAOCUOQ-UHFFFAOYSA-N
1,4-dipropylbenzene
C12H18
MFCD03844789
20958
CCCC1=CC=C(C=C1)CCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0025

TSCA : No

SDS
Documents

Documents

Promotions

Promotions