accessibility menu, dialog, popup

UserName

CH3CH3

CAS RN 4815-57-0

IUPAC Name: 1,4-dipropylbenzene
Synonyms: 1,4-dipropylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: PHUANMGFAOCUOQ-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(CCC)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results