missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dipropylbenzene 97.0+%, TCI America™

Catalog No. p-7127496
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27805ML 5 mL
D278025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27805ML Supplier TCI America Supplier No. D27805ML

Chemical Identifiers

CAS 4815-57-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD03844789
InChI Key PHUANMGFAOCUOQ-UHFFFAOYSA-N
PubChem CID 20958
IUPAC Name 1,4-dipropylbenzene
SMILES CCCC1=CC=C(C=C1)CCC

Specifications

Color Yellow
Boiling Point 221°C
Quantity 5 mL
Formula Weight 162.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dipropylbenzene
EINECSNumber (3)-0025
TSCA No