missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenylbutadiyne 98.0+%, TCI America™

Catalog No. D27895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27895G 5 g
D278925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27895G Supplier TCI America Supplier No. D27895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 886-66-8
Molecular Formula C16H10
Molecular Weight (g/mol) 202.26
MDL Number MFCD00004787
InChI Key HMQFJYLWNWIYKQ-UHFFFAOYSA-N
Synonym 1,4-diphenylbutadiyne, diphenylbutadiyne, diphenyldiacetylene, butadiyne, diphenyl, diphenyl-1,3-butadiyne, benzene, 1,1'-1,3-butadiyne-1,4-diyl bis, 1,4-diphenyl-1,3-butadiyne, 1,4-diphenylbuta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyldibenzene, 4-phenyl-1,3-butadiynyl benzene
PubChem CID 70174
ChEBI CHEBI:51588
IUPAC Name (4-phenylbuta-1,3-diyn-1-yl)benzene
SMILES C1=CC=C(C=C1)C#CC#CC1=CC=CC=C1

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 202.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenylbutadiyne