missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5%, Solstice

Catalog No. 6000079
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Narrow Mouth Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000079 4 x 2.5 L Glass Bottle
6000078 2.5 L Narrow Mouth Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000079 Supplier Solstice Supplier No. 348574X2.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ Plus, for HPLC,≥99.5%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 10°C to 12°C (literature)
Absorbance max. 0.01 at 295 nm, max. 0.10 at 270 nm, max. 0.20 at 250 nm, max. 0.50 at 235 nm, max. 0.70 at 220 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm
CAS Max % 100.0000%
Density 1.034 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point 11°C
Packaging Glass Bottle
Linear Formula C4H8O2
UN Number UN1165
Beilstein 102551
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
Boiling Point 100°C to 102°C
Molecular Weight (g/mol) 88.106
Formula Weight 88.11
Percent Purity ≥99.5%
Grade HPLC
Chemical Name or Material 1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5%
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Dioxane
EINECSNumber 204-661-8
Recommended Storage Room Temp