missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT

Catalog No. AA39118AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39118AP 500 mL
AA39118K2 1 L
AA39118AY 4 x 500 mL
AA39118K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA39118AP Supplier Thermo Scientific Chemicals Supplier No. 039118AP

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Furthermore, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide and in the preparation of histological sections for microscopic examination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 11.8°C
Color Colorless
Appearance Colorless liquid
Density 1.034 g/cm3 at 20°C
Quantity 500 mL
Flash Point 12°C (54°F)
Ignition Point 375°C (707°F)
Odor Ether like
Physical Form Liquid
Refractive Index 1.422
Assay Percent Range >99%
Vapor Pressure 41 hPa (31mm Hg) at 20°C
Beilstein 102551
Boiling Point 100°C to 102°C
Solubility Information Miscible with water,organic solvents,aromatic hydrocarbons and oils.
Formula Weight 88.11
Grade ACS
Concentration or Composition (by Analyte or Components) 1, 4-Dioxane
Chemical Name or Material 1, 4-Dioxane, For analysis ACS
Show More Show Less
Hazard Category H225-H319-H335-H336-H350-H373
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 204-661-8
RTECSNumber JG8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only