missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-2,3-diol 96.0+%, TCI America™

Catalog No. D206610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D206610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D206610G Supplier TCI America Supplier No. D206610G

Chemical Identifiers

CAS 4845-50-5
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.10
MDL Number MFCD00006572
InChI Key YLVACWCCJCZITJ-UHFFFAOYNA-N
Synonym 2,3-dihydroxy-1,4-dioxane, p-dioxane-2,3-diol, 2,3-dihydroxy-1,4-dioxan, dioxanediol, 1,3-diol, 1,4dioxane-2,3-diol, acmc-1al8g, dsstox_cid_18824, dsstox_rid_79404, dsstox_gsid_38824
PubChem CID 96170
IUPAC Name 1,4-dioxane-2,3-diol
SMILES OC1OCCOC1O

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 10 g
Physical Form Crystal-Powder at 20°C
Formula Weight 120.10
Percent Purity ≥96.0% (GC)
Chemical Name or Material 1,4-Dioxane-2,3-diol
EINECSNumber (5)-0831
TSCA Yes