missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dinitrobenzene 99.0+%, TCI America™

Numéro de catalogue. p-7126451
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D08195G 5 g
D081925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D08195G Fournisseur TCI America Code fournisseur D08195G
Il en reste null

Identifiants chimiques

CAS 100-25-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00007314
InChI Key FYFDQJRXFWGIBS-UHFFFAOYSA-N
Synonym p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
PubChem CID 7492
ChEBI CHEBI:51398
IUPAC Name 1,4-dinitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Spécifications

Melting Point 173°C
Quantity 5 g
UN Number 3443
Formula Weight 168.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dinitrobenzene
EINECSNumber (3)-0445
RTECSNumber CZ7525000
TSCA Yes