missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodobenzene 98.0+%, TCI America™

Catalog No. D06085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06085G 5 g
D060825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06085G Supplier TCI America Supplier No. D06085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-38-4
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.907
MDL Number MFCD00001054
InChI Key LFMWZTSOMGDDJU-UHFFFAOYSA-N
Synonym p-diiodobenzene, benzene, 1,4-diiodo, p-benzene diiodide, benzene, p-diiodo, unii-i9xcf83vav, i9xcf83vav, 1,4-diiodbenzene, 1,4-diidobenzene, 1,4 diiodobenzene, 1,4-diiodiobenzene
PubChem CID 12208
IUPAC Name 1,4-diiodobenzene
SMILES C1=CC(=CC=C1I)I

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 329.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diiodobenzene