missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodobenzene, 98%

Catalog No. AAA1533522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533522 100 g
AAA1533514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1533522 Supplier Thermo Scientific Chemicals Supplier No. A1533522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-38-4 | C6H4I2 | 329.907 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Ampouled under argon

Chemical Identifiers

CAS 624-38-4
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.907
MDL Number MFCD00001054
InChI Key LFMWZTSOMGDDJU-UHFFFAOYSA-N
Synonym p-diiodobenzene, benzene, 1,4-diiodo, p-benzene diiodide, benzene, p-diiodo, unii-i9xcf83vav, i9xcf83vav, 1,4-diiodbenzene, 1,4-diidobenzene, 1,4 diiodobenzene, 1,4-diiodiobenzene
PubChem CID 12208
IUPAC Name 1,4-diiodobenzene
SMILES C1=CC(=CC=C1I)I

Specifications

Melting Point 128°C to 132°C
Boiling Point 285°C
Quantity 100 g
Beilstein 1904546
Sensitivity Light sensitive
Solubility Information Soluble in ether and ethanol. Insoluble in water.
Formula Weight 329.91
Percent Purity 98%
Chemical Name or Material 1,4-Diiodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-842-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only