missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dihydroxynaphthalene 98.0+%, TCI America™
Supplier: TCI America D058925G
Specifications
| 1,4-Dihydroxynaphthalene | |
| 190°C | |
| MFCD00003977 | |
| 1,4-dihydroxynaphthalene, 1,4-naphthalenediol, 1,4-naphthohydroquinone, hydronaphthoquinone, naphthohydroquinone, 1,4-naphthoquinol, alpha-naphthoquinhydrone, unii-aml1p6t42c, ccris 7897, naphthalene-1,4-dio | |
| C1=CC=C2C(=C1)C(=CC=C2O)O | |
| 160.172 | |
| CHEBI:34063 | |
| ≥97.0% (GC,T) |
| 571-60-8 | |
| C10H8O2 | |
| 25 g | |
| PCILLCXFKWDRMK-UHFFFAOYSA-N | |
| naphthalene-1,4-diol | |
| 11305 | |
| 160.17 | |
| Crystalline Powder |
Chemical Identifiers
| 571-60-8 | |
| 160.172 | |
| PCILLCXFKWDRMK-UHFFFAOYSA-N | |
| 11305 | |
| naphthalene-1,4-diol |
| C10H8O2 | |
| MFCD00003977 | |
| 1,4-dihydroxynaphthalene, 1,4-naphthalenediol, 1,4-naphthohydroquinone, hydronaphthoquinone, naphthohydroquinone, 1,4-naphthoquinol, alpha-naphthoquinhydrone, unii-aml1p6t42c, ccris 7897, naphthalene-1,4-dio | |
| CHEBI:34063 | |
| C1=CC=C2C(=C1)C(=CC=C2O)O |
Safety and Handling
EINECSNumber : (4)-0356
RTECSNumber : QJ4730000
TSCA : Yes