accessibility menu, dialog, popup

UserName

OHOH

CAS RN 571-60-8

IUPAC Name: naphthalene-1,4-diol
Synonyms: 1,4-dihydroxynaphthalene α-naphthoquinhydrone 1,4-naphthoquinol hydronaphthoquinone 1,4-naphthohydroquinone
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: PCILLCXFKWDRMK-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results