missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxynaphthalene 98.0+%, TCI America™

Catalog No. p-7126304
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05895G 5 g
D058925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05895G Supplier TCI America Supplier No. D05895G

Chemical Identifiers

CAS 571-60-8
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00003977
InChI Key PCILLCXFKWDRMK-UHFFFAOYSA-N
Synonym 1,4-dihydroxynaphthalene, 1,4-naphthalenediol, 1,4-naphthohydroquinone, hydronaphthoquinone, naphthohydroquinone, 1,4-naphthoquinol, alpha-naphthoquinhydrone, unii-aml1p6t42c, ccris 7897, naphthalene-1,4-dio
PubChem CID 11305
ChEBI CHEBI:34063
IUPAC Name naphthalene-1,4-diol
SMILES C1=CC=C2C(=C1)C(=CC=C2O)O

Specifications

Melting Point 190°C
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dihydroxynaphthalene
EINECSNumber (4)-0356
RTECSNumber QJ4730000
TSCA Yes