missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diethoxybenzene 98.0+%, TCI America™
Supplier: TCI America D053525G
Specifications
| 1,4-Diethoxybenzene | |
| 70°C | |
| 112°C | |
| MFCD00015146 | |
| benzene, 1,4-diethoxy, p-diethoxybenzene, hydroquinone diethyl ether, benzene, p-diethoxy, diethoxybenzene, 1,4-diethoxy-benzen, benzene,4-diethoxy, 1,4-diethoxybenzol, hydroquinonedlethylether, acmc-209ans | |
| CCOC1=CC=C(OCC)C=C1 | |
| 166.22 | |
| 166.22 | |
| Crystalline Powder |
| 122-95-2 | |
| White-Yellow | |
| C10H14O2 | |
| 25 g | |
| VWGNFIQXBYRDCH-UHFFFAOYSA-N | |
| 1,4-diethoxybenzene | |
| 67150 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 122-95-2 | |
| 166.22 | |
| VWGNFIQXBYRDCH-UHFFFAOYSA-N | |
| 67150 | |
| CCOC1=CC=C(OCC)C=C1 |
| C10H14O2 | |
| MFCD00015146 | |
| benzene, 1,4-diethoxy, p-diethoxybenzene, hydroquinone diethyl ether, benzene, p-diethoxy, diethoxybenzene, 1,4-diethoxy-benzen, benzene,4-diethoxy, 1,4-diethoxybenzol, hydroquinonedlethylether, acmc-209ans | |
| 1,4-diethoxybenzene |
Safety and Handling
EINECSNumber : (3)-0582
TSCA : Yes