accessibility menu, dialog, popup

UserName

CAS RN 122-95-2

IUPAC Name: 1,4-diethoxybenzene
Synonyms: diethoxybenzene
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: VWGNFIQXBYRDCH-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(OCC)C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)