missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethoxybenzene 98.0+%, TCI America™

Catalog No. p-7126274
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D053525G 25 g
D0535500G 500 g
2 options

Catalog No. D053525G

Supplier: TCI America D053525G

Only null left

Chemical Identifiers

CAS 122-95-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015146
InChI Key VWGNFIQXBYRDCH-UHFFFAOYSA-N
Synonym benzene, 1,4-diethoxy, p-diethoxybenzene, hydroquinone diethyl ether, benzene, p-diethoxy, diethoxybenzene, 1,4-diethoxy-benzen, benzene,4-diethoxy, 1,4-diethoxybenzol, hydroquinonedlethylether, acmc-209ans
PubChem CID 67150
IUPAC Name 1,4-diethoxybenzene
SMILES CCOC1=CC=C(OCC)C=C1

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 112°C
Quantity 25 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diethoxybenzene
EINECSNumber (3)-0582
TSCA Yes