missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobutane, 97%

Catalog No. AAB2133730
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2133730 250 g
AAB213370B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2133730 Supplier Thermo Scientific Chemicals Supplier No. B2133730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-56-5 | C4H8Cl2 | 127.01 g/mol

Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.

Solubility
Soluble in water at 20°C2.4g/L.

Notes
Store away from oxidizing agents, alkali metals and alkali earth metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 110-56-5
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
MDL Number MFCD00001011
InChI Key KJDRSWPQXHESDQ-UHFFFAOYSA-N
Synonym butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
PubChem CID 8059
IUPAC Name 1,4-dichlorobutane
SMILES ClCCCCCl

Specifications

Melting Point -38°C
Density 1.139
Boiling Point 155°C
Flash Point 40°C (104°F)
Odor Aromatic
Refractive Index 1.454
Quantity 250 g
UN Number UN1993
Beilstein 635673
Solubility Information Soluble in water at 20°C2.4g/L.
Formula Weight 127.01
Percent Purity 97%
Chemical Name or Material 1,4-Dichlorobutane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-778-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only