missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibutoxybenzene 98.0+%, TCI America™

Catalog No. D32645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32645G Supplier TCI America Supplier No. D32645G
Only null left

Chemical Identifiers

CAS 104-36-9
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
InChI Key ZROGCHDPRZRKTI-UHFFFAOYSA-N
PubChem CID 66911
IUPAC Name 1,4-dibutoxybenzene
SMILES CCCCOC1=CC=C(C=C1)OCCCC

Specifications

Melting Point 45°C
Color White
Boiling Point 158°C
Quantity 5 g
Formula Weight 222.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibutoxybenzene