missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromotetrafluorobenzene 98.0+%, TCI America™
Supplier: TCI America D149325G
Specifications
| 1,4-Dibromotetrafluorobenzene | |
| 78°C | |
| C6Br2F4 | |
| 25 g | |
| QFTZULJNRAHOIY-UHFFFAOYSA-N | |
| 1,4-dibromo-2,3,5,6-tetrafluorobenzene | |
| 67653 | |
| ≥98.0% (GC) |
| 344-03-6 | |
| White-Yellow | |
| MFCD00000309 | |
| 1,4-dibromotetrafluorobenzene, p-dibromotetrafluorobenzene, benzene, 1,4-dibromo-2,3,5,6-tetrafluoro, benzene, 1,4-dibromotetrafluoro, 1,3,5,6-tetrafluorobenzene, benzene,4-dibromotetrafluoro, emolecules 536967, tetrafluoro-1,4-dibromo benzene, perfluoro-1,4-dibromobenzene | |
| FC1=C(F)C(Br)=C(F)C(F)=C1Br | |
| 307.87 | |
| 307.87 | |
| Crystalline Powder |
Chemical Identifiers
| 344-03-6 | |
| 307.87 | |
| QFTZULJNRAHOIY-UHFFFAOYSA-N | |
| 67653 | |
| FC1=C(F)C(Br)=C(F)C(F)=C1Br |
| C6Br2F4 | |
| MFCD00000309 | |
| 1,4-dibromotetrafluorobenzene, p-dibromotetrafluorobenzene, benzene, 1,4-dibromo-2,3,5,6-tetrafluoro, benzene, 1,4-dibromotetrafluoro, 1,3,5,6-tetrafluorobenzene, benzene,4-dibromotetrafluoro, emolecules 536967, tetrafluoro-1,4-dibromo benzene, perfluoro-1,4-dibromobenzene | |
| 1,4-dibromo-2,3,5,6-tetrafluorobenzene |
Safety and Handling
TSCA : No