accessibility menu, dialog, popup

UserName

FFBrFFBr

CAS RN 344-03-6

IUPAC Name: 1,4-dibromo-2,3,5,6-tetrafluorobenzene
Synonyms: 1,4-dibromo-2,3,5,6-tetrafluorobenzene
Molecular Weight (g/mol): 307.87
Molecular Formula: C6Br2F4
InChi Key: QFTZULJNRAHOIY-UHFFFAOYSA-N
SMILES: FC1=C(F)C(Br)=C(F)C(F)=C1Br
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)