missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromotetrafluorobenzene 98.0+%, TCI America™

Catalog No. p-7126749
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14935G 5 g
D149325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14935G Supplier TCI America Supplier No. D14935G

Chemical Identifiers

CAS 344-03-6
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.87
MDL Number MFCD00000309
InChI Key QFTZULJNRAHOIY-UHFFFAOYSA-N
Synonym 1,4-dibromotetrafluorobenzene, p-dibromotetrafluorobenzene, benzene, 1,4-dibromo-2,3,5,6-tetrafluoro, benzene, 1,4-dibromotetrafluoro, 1,3,5,6-tetrafluorobenzene, benzene,4-dibromotetrafluoro, emolecules 536967, tetrafluoro-1,4-dibromo benzene, perfluoro-1,4-dibromobenzene
PubChem CID 67653
IUPAC Name 1,4-dibromo-2,3,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(Br)=C(F)C(F)=C1Br

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 5 g
Formula Weight 307.87
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromotetrafluorobenzene