missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobenzene Zone Refined (number of passes:31) 99.8+%, TCI America™

Catalog No. D0297ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0297ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0297ZONE Supplier TCI America Supplier No. D02971SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-37-6
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000089
InChI Key SWJPEBQEEAHIGZ-UHFFFAOYSA-N
Synonym p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
PubChem CID 7804
ChEBI CHEBI:37150
IUPAC Name 1,4-dibromobenzene
SMILES BrC1=CC=C(Br)C=C1

Specifications

Melting Point 88°C
Color White
Boiling Point 219°C
Quantity ZONE
Formula Weight 235.91
Percent Purity ≥99.8% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromobenzene Zone Refined (number of passes:31)
EINECSNumber (3)-0052
RTECSNumber CZ1791000
TSCA Yes