missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromobenzene 99.0+%, TCI America™
Supplier: TCI America D0170100G
Specifications
| 1,4-Dibromobenzene | |
| 88°C | |
| 219°C | |
| MFCD00000089 | |
| p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071 | |
| BrC1=CC=C(Br)C=C1 | |
| 235.91 | |
| CHEBI:37150 | |
| ≥99.0% (GC) |
| 106-37-6 | |
| White-Yellow | |
| C6H4Br2 | |
| 100 g | |
| SWJPEBQEEAHIGZ-UHFFFAOYSA-N | |
| 1,4-dibromobenzene | |
| 7804 | |
| 235.91 | |
| Crystals |
Chemical Identifiers
| 106-37-6 | |
| 235.91 | |
| SWJPEBQEEAHIGZ-UHFFFAOYSA-N | |
| 7804 | |
| 1,4-dibromobenzene |
| C6H4Br2 | |
| MFCD00000089 | |
| p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071 | |
| CHEBI:37150 | |
| BrC1=CC=C(Br)C=C1 |
Safety and Handling
EINECSNumber : (3)-0052
RTECSNumber : CZ1791000
TSCA : Yes