accessibility menu, dialog, popup

UserName

CAS RN 106-37-6

IUPAC Name: 1,4-dibromobenzene
Synonyms: dibromobenzene C6H4br2
Molecular Weight (g/mol): 235.91
Molecular Formula: C6H4Br2
InChi Key: SWJPEBQEEAHIGZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)C=C1
Percent Purity 
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)
  • (4)
Physical Form 
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (7)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (4)
  • (1)
  • (6)