missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobenzene 99.0+%, TCI America™

Catalog No. p-7126070
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D017025G 25 g
D0170100G 100 g
D0170500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D017025G Supplier TCI America Supplier No. D017025G
Only null left

Chemical Identifiers

CAS 106-37-6
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000089
InChI Key SWJPEBQEEAHIGZ-UHFFFAOYSA-N
Synonym p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
PubChem CID 7804
ChEBI CHEBI:37150
IUPAC Name 1,4-dibromobenzene
SMILES BrC1=CC=C(Br)C=C1

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 235.91
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,4-Dibromobenzene
EINECSNumber (3)-0052
RTECSNumber CZ1791000
TSCA Yes